Thermo Scientific Chemicals

Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35, Thermo Scientific Chemicals

Catalog number: 228345000
500 g, HDPE plastic bottle, Each
Thermo Scientific Chemicals

Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35, Thermo Scientific Chemicals

Catalog number: 228345000
500 g, HDPE plastic bottle, Each
Quantity
Packaging
Catalog number: 228345000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9002-92-0
MDL Number
MFCD00043063
Infrared spectrum
Conforms
Hydroxyl value
40 to 60 mg KOH/g
Acid value
=<5 mg KOH/g
Impurity
=<10 ppm free Dioxane
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text