Chemical Identifiers
IUPAC Namebis[4-(dimethylamino)phenyl][4-(methylamino)phenyl]methylium
Molecular FormulaC24H28N3
InChI KeyHJPVVBCICYOONZ-UHFFFAOYSA-N
SMILESCNC1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
Molecular Weight (g/mol)358.51
View more