Thermo Scientific Chemicals

Methyl Violet 2B, pure, indicator grade

Catalog number: 228375000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl Violet 2B, pure, indicator grade

Catalog number: 228375000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 228375000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
8004-87-3
IUPAC Name
bis[4-(dimethylamino)phenyl][4-(methylamino)phenyl]methylium
Molecular Formula
C24H28N3
InChI Key
HJPVVBCICYOONZ-UHFFFAOYSA-N
SMILES
CNC1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
Appearance (Color)
Green
Appearance (Form)
Crystalline powder
Absorption Ratio P-15/P+15
1 to 1.25
Infrared spectrum
Conforms
UV spectrophotometry
>=75 % (on dry substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text