Thermo Scientific™

Pierce™ Midi Gel Power Staining Kit

Catalog number: 22839
Have Questions?
Thermo Scientific™

Pierce™ Midi Gel Power Staining Kit

Catalog number: 22839
Have Questions?
Catalog Number
22839
Unit Size
15 gels
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2283915 gels
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific™ Pierce™ Power Staining kits are designed for fast Coomassie dye staining of protein gels and optimized to work exclusively with the Pierce Power Stain Cassette and Pierce Power Station. The Pierce™ Midi Gel Power Staining Kit enables rapid Coomassie dye staining of proteins in midi-size polyacrylamide gels and the removal of unbound stain from the gel matrix.

• High performance—performance is equivalent to, or better than, traditional Coomassie dye staining techniques
• Fast—optimized ionic solutions enable Coomassie dye staining of protein and gel destaining within 6 to 11 minutes when used with the Pierce Power Stainer
• Compatible—verified to work with commonly used precast and homemade SDS-PAGE gels

Traditional Coomassie dye staining techniques require staining for one hour to overnight and prolonged destaining for desired results. When used in conjunction with Pierce Power Stain Solution, Destain Solution, and Gel Pads, the Pierce Power Stainer enables efficient staining and destaining in 6 to 11 minutes with results equivalent to, or better than, traditional Coomassie dye staining techniques.

Related Products
Pierce™ Mini Gel Power Staining Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
Midi Gel Power Staining Kit, 15 Gels, Coomassie Dye, Protein Gel Electrophoresis Application, Colorimetric Detection Method, In-Gel Detection, The Kit Contains Sufficient Reagents To Stain 15 Midi-Sized Gels at 30mL Each
Detection Location
In-Gel Detection
Detection Method
Colorimetric
Label or Dye
Coomassie
Product Type
Fast Protein Gel Stain
Formulation
Proprietary gel stain and destain solutions
Product Line
Pierce™
Quantity
1 Kit: 30 Pads, 15 Gels
Sufficient For
15 Midi-sized gels at 30mL each
Target Molecule
Protein

Contents & Storage

Note: The kit contains sufficient reagents to stain 15 midi-sized gels at 30 mL each.

Contents:
Midi Staining Gel Pads, 2 boxes of 15 each (8 sheets/pad)
Pierce Power Stain Solution, 480 mL
Destain Solution, 480 mL

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text