Thermo Scientific Chemicals

5-Bromo-2'-deoxyuridine, 99+%

Catalog number: 228590100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Bromo-2'-deoxyuridine, 99+%

Catalog number: 228590100
10 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 228590100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59-14-3
IUPAC Name
5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Formula
C9H11BrN2O5
InChI Key
WOVKYSAHUYNSMH-RRKCRQDMSA-N
SMILES
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O
Related substances
=<0.5 % (HPLC) 5-Bromouracil
Related substances
=<0.2 % (HPLC) Each unknown impurity
Related substances
=<1.0 % (HPLC) Total impurities
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text