Thermo Scientific Chemicals

D(+)-Proline, ≥99%

Catalog number: 228660250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

D(+)-Proline, ≥99%

Catalog number: 228660250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 228660250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
344-25-2
IUPAC Name
pyrrolidine-2-carboxylic acid
Molecular Formula
C5H9NO2
InChI Key
ONIBWKKTOPOVIA-UHFFFAOYNA-N
SMILES
OC(=O)C1CCCN1
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with HClO4
99.0 to 101.0 %
Solubility
(5 % in water) Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text