Thermo Scientific Chemicals

Chloroxazone, 98%, Thermo Scientific Chemicals

Catalog number: 228950250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Chloroxazone, 98%, Thermo Scientific Chemicals

Catalog number: 228950250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 228950250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
95-25-0
IUPAC Name
5-chloro-2,3-dihydro-1,3-benzoxazol-2-one
Molecular Formula
C7H4ClNO2
InChI Key
TZFWDZFKRBELIQ-UHFFFAOYSA-N
SMILES
ClC1=CC=C2OC(=O)NC2=C1
Appearance (Color)
White to yellow-brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
189°C to 194°C
Titration with NaOH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text