Urocanic acid, 98%, Thermo Scientific Chemicals
Urocanic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Urocanic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 228960050
Price (USD)/ Each
45.65
Online exclusive
51.10 
Save 5.45 (11%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
45.65
Online exclusive
51.10 
Save 5.45 (11%)
Add to cart
Urocanic acid, 98%, Thermo Scientific Chemicals
Catalog number228960050
Price (USD)/ Each
45.65
Online exclusive
51.10 
Save 5.45 (11%)
-
Add to cart
Chemical Identifiers
CAS104-98-3
IUPAC Name(2E)-3-(1H-imidazol-5-yl)prop-2-enoic acid
Molecular FormulaC6H6N2O2
InChI KeyLOIYMIARKYCTBW-OWOJBTEDSA-N
SMILESOC(=O)C=CC1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder
Infrared spectrumConforms
Melting point217°C to 220°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only