Thermo Scientific Chemicals

Urocanic acid, 98%, Thermo Scientific Chemicals

Catalog number: 228960250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Urocanic acid, 98%, Thermo Scientific Chemicals

Catalog number: 228960250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 228960250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
104-98-3
IUPAC Name
(2E)-3-(1H-imidazol-5-yl)prop-2-enoic acid
Molecular Formula
C6H6N2O2
InChI Key
LOIYMIARKYCTBW-OWOJBTEDSA-N
SMILES
OC(=O)C=CC1=CN=CN1
Appearance (Color)
White to beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
217°C to 220°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text