Thermo Scientific Chemicals

L-Valine methyl ester hydrochloride, 99%

Catalog number: 229260100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

L-Valine methyl ester hydrochloride, 99%

Catalog number: 229260100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 229260100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6306-52-1
IUPAC Name
methyl 2-amino-3-methylbutanoate hydrochloridyl
Molecular Formula
C6H13ClNO2
InChI Key
RFMIEHOYULVMNZ-UHFFFAOYNA-N
SMILES
[Cl].COC(=O)C(N)C(C)C
Titration Argentometric
>=98.5 %
Solubility
(5 % in water) Clear colorless to light yellow
Appearance (Form)
Powder or crystals
Appearance (Color)
White to off-white
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text