L-Valine methyl ester hydrochloride, 99%
L-Valine methyl ester hydrochloride, 99%
Thermo Scientific Chemicals

L-Valine methyl ester hydrochloride, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 229260500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namemethyl 2-amino-3-methylbutanoate hydrochloridyl
Molecular FormulaC6H13ClNO2
InChI KeyRFMIEHOYULVMNZ-UHFFFAOYNA-N
SMILES[Cl].COC(=O)C(N)C(C)C
Molecular Weight (g/mol)166.63
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=98.5 %
Solubility(5 % in water) Clear colorless to light yellow
Appearance (Form)Powder or crystals
Appearance (Color)White to off-white
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only