Thermo Scientific Chemicals

Gluconic acid, potassium salt, 99%, Thermo Scientific Chemicals

Catalog number: 229322500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Gluconic acid, potassium salt, 99%, Thermo Scientific Chemicals

Catalog number: 229322500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 229322500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
299-27-4
IUPAC Name
potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
Molecular Formula
C6H11KO7
InChI Key
HLCFGWHYROZGBI-JJKGCWMISA-M
SMILES
[K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
Appearance (Color)
White to off-white
Loss on drying
=<3 % (105°C, 4 h) (Vacuum)
Infrared spectrum
Conforms
Reducing substances
=<1 % (as Dextrose)
Appearance (Form)
Crystalline or granular powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text