Thermo Scientific Chemicals

L-Methionine methyl ester hydrochloride, 98%

Catalog number: 229350500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

L-Methionine methyl ester hydrochloride, 98%

Catalog number: 229350500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 229350500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2491-18-1
IUPAC Name
hydrogen methyl 2-amino-4-(methylsulfanyl)butanoate chloride
Molecular Formula
C6H14ClNO2S
InChI Key
MEVUPUNLVKELNV-UHFFFAOYNA-N
SMILES
[H+].[Cl-].COC(=O)C(N)CCSC
Titration Argentometric
97.5 to 102.5 %
Solubility
(5 % in methanol) Clear colorless to light yellow
Infrared spectrum
Conforms
Appearance (Color)
White to beige
Specific optical rotation
+24° to +28° (20°C, 589 nm) (c=1, water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text