Salicylamide, 99%
Salicylamide, 99%
Salicylamide, 99%
Thermo Scientific Chemicals

Salicylamide, 99%

CAS: 65-45-2 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
Catalog number 229362500
Price (USD)
92.50
Each
In stock
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
92.50
Each
Add to cart
Chemical Identifiers
CAS65-45-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point139°C to 142°C
Water=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link