Thermo Scientific Chemicals

Myristyltrimethylammonium bromide, 99%, Thermo Scientific Chemicals

Catalog number: 229370025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Myristyltrimethylammonium bromide, 99%, Thermo Scientific Chemicals

Catalog number: 229370025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 229370025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1119-97-7
IUPAC Name
trimethyl(tetradecyl)azanium bromide
Molecular Formula
C17H38BrN
InChI Key
CXRFDZFCGOPDTD-UHFFFAOYSA-M
SMILES
[Br-].CCCCCCCCCCCCCC[N+](C)(C)C
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
245°C to 250°C
Appearance (Color)
White
Titration Argentometric
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text