Thermo Scientific Chemicals

Polyvinylpyrrolidone-iodine complex, Thermo Scientific Chemicals

Catalog number: 229470025
2.5 kg, Each
Thermo Scientific Chemicals

Polyvinylpyrrolidone-iodine complex, Thermo Scientific Chemicals

Catalog number: 229470025
2.5 kg, Each
Quantity
Catalog number: 229470025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25655-41-8
Molecular Formula
(C6H9NO)nI2
SMILES
II.*-CC(-*)N1CCCC1=O
MDL Number
MFCD00084483
Appearance (Form)
Powder
Assay
9.0 to 12.0 % (available iodine)
Heavy metals (as Pb)
=<20 ppm
Nitrogen (N)
9.5 to 11.5 % (on dried substance)
Appearance (Color)
Yellow to red-brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text