Polyvinylpyrrolidone-iodine complex, Thermo Scientific Chemicals
Polyvinylpyrrolidone-iodine complex, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyvinylpyrrolidone-iodine complex, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Catalog number 229475000
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
Molecular Formula(C6H9NO)nI2
SMILESII.*-CC(-*)N1CCCC1=O
MDL NumberMFCD00084483
CAS25655-41-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Assay9.0 to 12.0 % (available iodine)
Heavy metals (as Pb)=<20 ppm
Nitrogen (N)9.5 to 11.5 % (on dried substance)
Appearance (Color)Yellow to red-brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only