Thermo Scientific Chemicals

Brilliant cresyl Blue, pure, high purity biological stain

Catalog number: 229590100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Brilliant cresyl Blue, pure, high purity biological stain

Catalog number: 229590100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 229590100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81029-05-2
IUPAC Name
7,9-diamino-N,N-diethyl-6-methyl-5a,9a-dihydro-3H-phenoxazin-3-iminium chloride
Molecular Formula
C17H23ClN4O
InChI Key
SHMCAAFKAHAURK-UHFFFAOYNA-M
SMILES
[Cl-].CC[N+](CC)=C1C=CC2=NC3C(OC2=C1)C(C)=C(N)C=C3N
Appearance (Color)
Green to dark blue
Infrared spectrum
Conforms
Solubility
(0.1% in 50% ethanol) Clear blue
Specific absorbance A (1%/1cm)
>=1500 (625 to 630 nm, 50% ethanol)
Loss on drying
=<8 % (1 g, 110°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text