Chemical Identifiers
IUPAC Name7,9-diamino-N,N-diethyl-6-methyl-5a,9a-dihydro-3H-phenoxazin-3-iminium chloride
Molecular FormulaC17H23ClN4O
InChI KeySHMCAAFKAHAURK-UHFFFAOYNA-M
SMILES[Cl-].CC[N+](CC)=C1C=CC2=NC3C(OC2=C1)C(C)=C(N)C=C3N
Molecular Weight (g/mol)334.85
View more