Thermo Scientific Chemicals

Brilliant Green, pure, high purity biological stain

Catalog number: 229601000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Brilliant Green, pure, high purity biological stain

Catalog number: 229601000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 229601000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
633-03-4
IUPAC Name
bis[4-(diethylamino)phenyl](phenyl)methylium hydrogen sulfate
Molecular Formula
C27H34N2O4S
InChI Key
NNBFNNNWANBMTI-UHFFFAOYSA-M
SMILES
OS([O-])(=O)=O.CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(CC)CC
Appearance (Color)
Green to green-yellow
UV Lambda max
625 to 631 nm (in 50% ethanol)
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Loss on drying
=<5 % (105°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text