Thermo Scientific Chemicals

Bromocresol Purple, sodium salt, pure, water soluble, indicator

Catalog number: 229610250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Bromocresol Purple, sodium salt, pure, water soluble, indicator

Catalog number: 229610250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 229610250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62625-30-3
IUPAC Name
2-[(3-bromo-4-hydroxy-5-methylphenyl)[(1E)-3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
Molecular Formula
C21H15Br2O5S
InChI Key
HJZFRHKAXRLQSG-XMHGGMMESA-M
SMILES
CC1=CC(=CC(Br)=C1O)C(=C1\C=C(C)C(=O)C(Br)=C1)\C1=CC=CC=C1S([O-])(=O)=O
Appearance (Form)
Powder
Specific extinction E (1%/1cm)
>=900 (590 nm, 0.01 N NaOH)
Appearance (Color)
Red-brown
Infrared spectrum
Conforms
Solubility
(0.1 % in water) Clear

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text