Bromocresol Purple, sodium salt, pure, water soluble, indicator
Bromocresol Purple, sodium salt, pure, water soluble, indicator
Thermo Scientific Chemicals

Bromocresol Purple, sodium salt, pure, water soluble, indicator

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
10 g
25 g
Packaging:
Glass bottle
Catalog number 229611000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS62625-30-3
IUPAC Name2-[(3-bromo-4-hydroxy-5-methylphenyl)[(1E)-3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
Molecular FormulaC21H15Br2O5S
InChI KeyHJZFRHKAXRLQSG-XMHGGMMESA-M
SMILESCC1=CC(=CC(Br)=C1O)C(=C1\C=C(C)C(=O)C(Br)=C1)\C1=CC=CC=C1S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Specific extinction E (1%/1cm)>=900 (590 nm, 0.01 N NaOH)
Appearance (Color)Red-brown
Infrared spectrumConforms
Solubility(0.1 % in water) Clear
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only