Cresyl Violet acetate, pure, high purity biological stain
Cresyl Violet acetate, pure, high purity biological stain
Cresyl Violet acetate, pure, high purity biological stain
Cresyl Violet acetate, pure, high purity biological stain
Thermo Scientific Chemicals

Cresyl Violet acetate, pure, high purity biological stain

CAS: 10510-54-0 | C18H15N3O3 | 321.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 229630050
Price (USD)
98.65
Special offer
Online exclusive
Ends: 26-Sep-2025
122.00
Save 23.35 (19%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
98.65
Special offer
Online exclusive
Ends: 26-Sep-2025
122.00
Save 23.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS10510-54-0
IUPAC Name8,12-diamino-10λ⁴-oxa-5-azatetraphen-10-ylium acetate
Molecular FormulaC18H15N3O3
InChI KeyXKOCOMKJPWEOHX-UHFFFAOYSA-M
SMILESCC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3
View more
SpecificationsSpecification SheetSpecification Sheet
Dye content>=65 %
Appearance (Color)Dark green to black
Infrared spectrumConforms
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection