Crystal Violet, pure, high purity biological stain
Crystal Violet, pure, high purity biological stain
Crystal Violet, pure, high purity biological stain
Crystal Violet, pure, high purity biological stain
Thermo Scientific Chemicals

Crystal Violet, pure, high purity biological stain

CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 229641000
Price (USD)
101.65
Online Exclusive
113.00
Save 11.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
101.65
Online Exclusive
113.00
Save 11.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS548-62-9
IUPAC Nametris[4-(dimethylamino)phenyl]methylium chloride
Molecular FormulaC25H30ClN3
InChI KeyZXJXZNDDNMQXFV-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to green-brown
Infrared spectrumConforms
Loss on drying=<9 % (110°C)
Sulfated ash=<3 %
Insoluble matter(in ethanol) Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Share catalog number, name or link