Eosin B, pure, high purity, biological stain
Eosin B, pure, high purity, biological stain
Eosin B, pure, high purity, biological stain
Eosin B, pure, high purity, biological stain
Thermo Scientific Chemicals

Eosin B, pure, high purity, biological stain

Eosin B, C20H6Br2N2Na2O9, CAS Number-548-24-3, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, 4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, Eosin Bluish, 100g, 0.96 to 1.22, 19, 233, Brown, 208-943-1, 624.06, Pure, Danger, GHS H Statement: Causes serious eye damage. | CAS: 548-24-3 | C20H6Br2N2Na2O9 | 624.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 229650250
Price (USD)
36.65
Online Exclusive
40.40
Save 3.75 (9%)
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
36.65
Online Exclusive
40.40
Save 3.75 (9%)
Each
Add to cart
Chemical Identifiers
CAS548-24-3
IUPAC Namedisodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular FormulaC20H6Br2N2Na2O9
InChI KeyGYYTYUGGVBYJHE-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(C=C12)[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
UV Lambda max511 to 520 nm (in water + 1% Na2CO3)
Absorption Ratio P-15/P+150.96 to 1.22
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link