Thermo Scientific Chemicals

Eosin B, pure, high purity, biological stain

Catalog number: 229651000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Eosin B, pure, high purity, biological stain

Catalog number: 229651000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 229651000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
548-24-3
IUPAC Name
disodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular Formula
C20H6Br2N2Na2O9
InChI Key
GYYTYUGGVBYJHE-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(C=C12)[N+]([O-])=O)[N+]([O-])=O
Appearance (Color)
Dark brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
UV Lambda max
511 to 520 nm (in water + 1% Na2CO3)
Absorption Ratio P-15/P+15
0.96 to 1.22

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text