Thermo Scientific Chemicals

Erioglaucine disodium salt, pure

Catalog number: 229730250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Erioglaucine disodium salt, pure

Catalog number: 229730250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 229730250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3844-45-9
IUPAC Name
disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(2-sulfonatophenyl)methylium
Molecular Formula
C37H34N2Na2O9S3
InChI Key
SGHZXLIDFTYFHQ-UHFFFAOYSA-L
SMILES
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=CC=C1S([O-])(=O)=O
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
UV spectrum
Complies
Loss on drying
=<10 % (105°C)
UV Lambda max
625 to 630 nm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text