Thermo Scientific Chemicals

Light Green SF, Yellowish, pure, high purity biological stain

Catalog number: 229770100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Light Green SF, Yellowish, pure, high purity biological stain

Catalog number: 229770100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 229770100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5141-20-8
IUPAC Name
disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-sulfonatophenyl)methylium
Molecular Formula
C37H34N2Na2O9S3
InChI Key
DGOBMKYRQHEFGQ-UHFFFAOYSA-L
SMILES
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S([O-])(=O)=O
Appearance (Color)
Purple
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Titration
>=60 % (As is)
Loss on drying
=<10 % (1 g, 110°C, 2 h)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text