Chemical Identifiers
CAS5141-20-8
IUPAC Namedisodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-sulfonatophenyl)methylium
Molecular FormulaC37H34N2Na2O9S3
InChI KeyDGOBMKYRQHEFGQ-UHFFFAOYSA-L
SMILES[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S([O-])(=O)=O
View more