Thermo Scientific Chemicals

Malachite Green oxalate, pure, high purity biological stain

Catalog number: 229785000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Malachite Green oxalate, pure, high purity biological stain

Catalog number: 229785000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 229785000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2437-29-8
IUPAC Name
bis(bis[4-(dimethylamino)phenyl](phenyl)methylium) oxalic acid dihydrogen oxalate
Molecular Formula
C52H54N4O12
InChI Key
CNYGFPPAGUCRIC-UHFFFAOYSA-L
SMILES
OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
UV Lambda max
615 to 619 nm (water)
Absorption Ratio P-15/P+15
0.9 to 1.15
Specific absorbance A (1%/1cm)
>=1800 (at lambda max.)
Infrared spectrum
Conforms
Loss on drying
=<10 % (105°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text