Methylene Blue hydrate, 96+%, high purity biological stain
Methylene Blue hydrate, 96+%, high purity biological stain
Thermo Scientific Chemicals

Methylene Blue hydrate, 96+%, high purity biological stain

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Catalog number 229801000
Price (USD)/ Each
67.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
79.30 
Save 11.65 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
67.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
79.30 
Save 11.65 (15%)
Add to cart
Methylene Blue hydrate, 96+%, high purity biological stain
Catalog number229801000
Price (USD)/ Each
67.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
79.30 
Save 11.65 (15%)
-
Add to cart
Chemical Identifiers
CAS122965-43-9
IUPAC Name7-(dimethylamino)-N,N-dimethyl-3H-phenothiazin-3-iminium chloride
Molecular FormulaC16H18ClN3S
InChI KeyCXKWCBBOMKCUKX-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Dye content96.0 to 103.0 % (on dried substance)
Loss on drying8.0 % to 22.0 % (105°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only