Chemical Identifiers
CAS1936-15-8
IUPAC Namedisodium (8Z)-7-oxo-8-(2-phenylhydrazin-1-ylidene)-7,8-dihydronaphthalene-1,3-disulfonate
Molecular FormulaC16H10N2Na2O7S2
InChI KeyAEOVEGJBKQQFOP-DDVLFWKVSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O
View more