Thermo Scientific Chemicals

Thionin acetate, pure, high purity biological stain

Catalog number: 229840050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Thionin acetate, pure, high purity biological stain

Catalog number: 229840050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 229840050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78338-22-4
IUPAC Name
3,7-diamino-5λ⁴-phenothiazin-5-ylium
Molecular Formula
C12H10N3S
InChI Key
CBBMGBGDIPJEMI-UHFFFAOYSA-N
SMILES
NC1=CC2=[S+]C3=CC(N)=CC=C3N=C2C=C1
UV spectrum
Complies
Appearance (Color)
Dark green
Appearance (Form)
Crystalline powder or crystals
Dye content
>=85 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text