Thermo Scientific Chemicals

4,5',8-Trimethylpsoralen, 97.5%, Thermo Scientific Chemicals

Catalog number: 229881000
100 mg, Each
Thermo Scientific Chemicals

4,5',8-Trimethylpsoralen, 97.5%, Thermo Scientific Chemicals

Catalog number: 229881000
100 mg, Each
Quantity
Catalog number: 229881000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3902-71-4
IUPAC Name
2,5,9-trimethyl-7H-furo[3,2-g]chromen-7-one
Molecular Formula
C14H12O3
InChI Key
FMHHVULEAZTJMA-UHFFFAOYSA-N
SMILES
CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1
HPLC
>=96.5 % (on dried substance)
Melting point
230°C to 237°C
Loss on drying
=<0.5 % (105°C, 6 hrs)
Appearance (Color)
White to light beige
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text