Thermo Scientific Chemicals

3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 229990050
5 g, Each
Thermo Scientific Chemicals

3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 229990050
5 g, Each
Quantity
Catalog number: 229990050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
207738-08-7
IUPAC Name
3,3',5,5'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine dihydrochloride
Molecular Formula
C16H22Cl2N2
InChI Key
NYNRGZULARUZCC-UHFFFAOYSA-N
SMILES
Cl.Cl.CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1
Appearance (Color)
Off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration Argentometric
>=98.0 % (on dry substance)
Water
=<12.0 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text