VersaRack for VersaCool™ Refrigerated Circulating Bath, Stainless steel, adjustable
VersaRack for VersaCool™ Refrigerated Circulating Bath, Stainless steel, adjustable
Thermo Scientific™
VersaRack for VersaCool™ Refrigerated Circulating Bath, Stainless steel, adjustable
Stainless steel adjustable rack. For use with Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
Have Questions?
Catalog number 230000001
Price (USD)/ Each
-
Request A Quote
Description:
VersaRack - Stainless steel adjustable rack
Utilize the VersaRack for optimal performance with the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
With the ability to accommodate a wide range of beaker and test tube sizes, the VersaRack offers a
tool-less adjustable height setting . Its drip-less features allows it to rests over the bath opening saving you valuable bench space.
Specifications
DescriptionVersaRack - Stainless steel adjustable rack
For Use With (Equipment)Thermo Scientific VersaCool Refrigerated Circulating Bath
TypeTube Rack
HoldsHolds up to 75 test Tubes that are 10 mm dia.
Holds up to 48 test Tubes that are 16 mm dia.
Holds up to 21 test Tubes that are 25 mm dia.
Unit SizeEach