Thermo Scientific Chemicals

D(+)-Biotin, 98%

Catalog number: 230090050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

D(+)-Biotin, 98%

Catalog number: 230090050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 230090050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58-85-5
IUPAC Name
5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
Molecular Formula
C10H16N2O3S
InChI Key
YBJHBAHKTGYVGT-UHFFFAOYNA-N
SMILES
OC(=O)CCCCC1SCC2NC(=O)NC12
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 % (On dried substance)
Specific optical rotation
+89° to +93° (20°C, 589 nm) (c=2, 0.1 N NaOH)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text