Thermo Scientific™

SPB (succinimidyl-[4-(psoralen-8-yloxy)]-butyrate)

Catalog number: 23013
Have Questions?
Thermo Scientific™

SPB (succinimidyl-[4-(psoralen-8-yloxy)]-butyrate)

Catalog number: 23013
Have Questions?
Catalog Number
23013
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2301350 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce SPB is an NHS-ester and psoralen crosslinker that effectively conjugates primary amines to DNA via light-activated intercalation next to pyrimidine bases.

Features of SPB:

Reactive groups: NHS ester and psoralen
Reactive toward: amino groups and thymine
• DNA intercalating agent and nucleic acid probe1,2
• Reagent for photo-immobilization to microplates3
• Melting point: 177-178°C

Product References:
Crosslinker Application Guide -- search for recent literature references for this product
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Amine-Nonselective
Cleavable
No
Description
SPB
Molecular Weight (g/mol)
385.32
PEGylated
No
Spacer Arm Length
8.6 Å
Cell Permeability
Not determined
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Heterobifunctional
Reactive Moiety
Psoralen, NHS Ester
Spacer
Short (<10 Å)
Form
Solid
Quantity
50 mg
Solubility
DMF, DMSO
Format
Standard
Water Soluble
No
Product Type
Crosslinker

Contents & Storage

Upon receipt store desiccated at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text