Laminin is the major glycoprotein component of basement membranes, and has a molecular weight of 850,000 daltons. This preparation ofRead more
Have Questions?
Catalog Number
Quantity
23017015
1 mg
Catalog number 23017015
Price (USD)
264.65
Online Exclusive
286.00
Save 21.35 (7%)
Each
-
Add to cart
Quantity:
1 mg
Price (USD)
264.65
Online Exclusive
286.00
Save 21.35 (7%)
Each
Add to cart
Laminin is the major glycoprotein component of basement membranes, and has a molecular weight of 850,000 daltons. This preparation of mouse Laminin was isolated from the Engelbreth-Holm-Swarm (EHS) sarcoma and is supplied at a concentration of 0.5-2.0 mg⁄ml.
Key functions include:
• Modulation of cell attachment, cell spreading, cell growth and motility • Binding to itself and other matrix compounds • Promotion of epithelial differentiation • Modification of leukocyte function • Stimulation of neurite outgrowth
Supplied: 1 mg in 50 mM Tris-HCL (pH 7.4), 0.15 M NaCl
Performance qualification (cell attachment): Half-maximal cell attachment of MG63 cells at 37°C: ≤ 20 ug⁄ml
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gene AliasLaminin
Protein SubtypeLaminin
Quantity1 mg
Research CategoryExtracellular Matrix & Cell Adhesion, Stem Cell Research
Shipping ConditionDry Ice
For Use With (Application)Cell Culture
FormLiquid
SpeciesMouse
Unit SizeEach
Contents & Storage
Store in ultra-cold freezer (-68 to -85°C) and protect from light.
Please enter sku to load default questions
Frequently asked questions (FAQs)
What is the molecular weight of Laminin Mouse Protein, Natural?
It has 3 subunits and has a MW of 850 kDa: one A chain is 400 kDa and two B chains are 205 and 215 kDa.
What is the protein concentration per vial for natural mouse laminin?
The protein concentration per vial is 1 mg/vial. The specification for protein concentration can range from 0.5 to 2.0 mg/mL. The volume per vial can vary as it will be based on the protein concentration determined for the specific lot. The volume added to each vial will equal 1 mg of laminin.