Thermo Scientific Chemicals

L(-)-Carnitine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 230281000
100 g, Each
Thermo Scientific Chemicals

L(-)-Carnitine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 230281000
100 g, Each
Quantity
Catalog number: 230281000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6645-46-1
IUPAC Name
[(2R)-3-carboxy-2-hydroxypropyl]trimethylazanium chloride
Molecular Formula
C7H16ClNO3
InChI Key
JXXCENBLGFBQJM-FYZOBXCZSA-N
SMILES
[Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
97.5 to 102.5 % (On dry substance)
Specific optical rotation
-21° to -24° (20°C, 589 nm) (c=1, H2O)
Sulfated ash
=<0.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text