Dodecyl sulfate, sodium salt, 99%, for biochemistry
Dodecyl sulfate, sodium salt, 99%, for biochemistry
Dodecyl sulfate, sodium salt, 99%, for biochemistry
Dodecyl sulfate, sodium salt, 99%, for biochemistry
Thermo Scientific Chemicals

Dodecyl sulfate, sodium salt, 99%, for biochemistry

CAS: 151-21-3 | C12H25NaO4S, C12H25NaO4S | 288.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
100 g
500 g
2.5 kg
Catalog number 230420025
Price (EUR)
-
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS151-21-3
IUPAC Namesodium dodecyl sulfate
Molecular FormulaC12H25NaO4S
InChI KeyDBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<1 % (1 g, 105°C, 3 h)
UV(3 % in water)
Solubility(3 % in water) Clear colorless
Chloride (Cl)=<500 ppm
UVat 260 nm A: =<0.05
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0284596Certificate of AnalysisMar 27, 2025230425000, 230421000, 230420025, 230420100, 230420250
A0468577Certificate of AnalysisJan 08, 2025230425000, 230421000, 230420025, 230420100, 230420250
B0155914ACertificate of AnalysisNov 26, 2024230425000, 230421000, 230420025, 230420100, 230420250
A0464867Certificate of AnalysisAug 07, 2024230425000, 230421000, 230420025, 230420100, 230420250
A0462201Certificate of AnalysisJun 24, 2024230425000, 230421000, 230420025, 230420100, 230420250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable solids
Category 2
Acute oral toxicity
Category 4
Acute Inhalation Toxicity - Dusts and Mists
Category 4
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 1
Specific target organ toxicity - (single exposure)
Category 3
Chronic aquatic toxicity
Category 3
Label Elements
Signal Word

Danger

Hazard Statements

H302 + H332 - Harmful if swallowed or if inhaled

H315 - Causes skin irritation

H318 - Causes serious eye damage

H335 - May cause respiratory irritation

H412 - Harmful to aquatic life with long lasting effects

Physical Hazards

H228 - Flammable solid

EU Specific Hazard Statements

May form combustible dust concentrations in air

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Share catalog number, name or link

1x1 image pixel for data collection