Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

Dodecyl sulfate, sodium salt, 99%, for biochemistry

Catalog number: 230420100
10 g, Each
Thermo Scientific Chemicals

Dodecyl sulfate, sodium salt, 99%, for biochemistry

Catalog number: 230420100
10 g, Each
Quantity
Catalog number: 230420100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
151-21-3
IUPAC Name
sodium dodecyl sulfate
Molecular Formula
C12H25NaO4S
InChI Key
DBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Chloride (Cl)
=<500 ppm
UV
at 260 nm A: =<0.05
Infrared spectrum
Conforms
Loss on drying
=<1 % (1 g, 105°C, 3 h)
UV
(3 % in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text