Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Prepare large volume media and cultures with Thermo Scientific™ Nalgene™ Round Polycarbonate Clearboy™ with Spigot. These durable Clearboy are lighter and safer than glass, and especially useful for visual inspection of contents for quality. Ideal for refrigerated or frozen storage of aqueous solutions, or for dispensing sterile water, aqueous reagents and media. Nalgene polycarbonate carboys can be autoclaved before refilling to eliminate bacterial growth when storing sterile solutions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Autoclavable | Autoclavable |
Capacity (English) | 5.3 gal. |
Capacity (Metric) | 20 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | Clear |
Description | Transparent Carboy with Spigot PC 20 L |
Diameter (Metric) Inner Neck | 66 mm |
Diameter (Metric) Outer | 290 mm |
For Use With (Application) | Suitable for mixing, storing and freezing aqueous-based reagents and biologicals, especially useful for visual inspection of contents for quality. |
Graduations | Molded-in graduations in 1 gal. and 5 L increments |
Height (Metric) With Closure | 536 mm |
Includes | Spigot |
Material | Polycarbonate |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Spigot |
Unit Size | Case of 4 |
Autoclavable | Autoclavable |
Capacity (English) | 2.6 gal. |
Capacity (Metric) | 10 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | Clear |
Description | Transparent Carboy with Spigot PC 10 L |
Diameter (Metric) Inner Neck | 66 mm |
Diameter (Metric) Outer | 253 mm |
For Use With (Application) | Suitable for mixing, storing and freezing aqueous-based reagents and biologicals, especially useful for visual inspection of contents for quality. |
Graduations | Molded-in graduations in 1 gal. and 5 L increments |
Height (Metric) With Closure | 394 mm |
Includes | Spigot |
Material | Polycarbonate |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Spigot |
Unit Size | Case of 4 |
Autoclavable | Autoclavable |
Capacity (English) | 2.6 gal. |
Capacity (Metric) | 10 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | Clear |
Description | Transparent Carboy with Spigot PC 10 L |
Diameter (Metric) Inner Neck | 66 mm |
Diameter (Metric) Outer | 253 mm |
For Use With (Application) | Suitable for mixing, storing and freezing aqueous-based reagents and biologicals, especially useful for visual inspection of contents for quality. |
Graduations | Molded-in graduations in 1 gal. and 5 L increments |
Height (Metric) With Closure | 394 mm |
Includes | Spigot |
Material | Polycarbonate |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Spigot |
Unit Size | Each |
Autoclavable | Autoclavable |
Capacity (English) | 5.3 gal. |
Capacity (Metric) | 20 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | Clear |
Description | Transparent Carboy with Spigot PC 20 L |
Diameter (Metric) Inner Neck | 66 mm |
Diameter (Metric) Outer | 290 mm |
For Use With (Application) | Suitable for mixing, storing and freezing aqueous-based reagents and biologicals, especially useful for visual inspection of contents for quality. |
Graduations | Molded-in graduations in 1 gal. and 5 L increments |
Height (Metric) With Closure | 536 mm |
Includes | Spigot |
Material | Polycarbonate |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Spigot |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2317-0050 | Case of 4 | 20 L | 1,594.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2317-0020 | Case of 4 | 10 L | 958.19 Online price 1,234.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2317-0020PK also known as 2317-0020 | Each | 10 L | 364.30 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2317-0050PK also known as 2317-0050 | Each | 20 L | 470.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Includes:
Polypropylene spigot screw closure and TPE gasket. See autoclaving instructions for more information.
Note:
Completely disengage closure threads and disassemble spigot before autoclaving
Follow product insert instructions for autoclaving to ensure proper function
Note: Tubing adaptor is not autoclavable
Repeated autoclaving will shorten the life of polycarbonate carboys
Use pH neutral detergent and rinse completely prior to autoclaving
Please recycle the corrugate cartons and LDPE bags in which your Nalgene carboy products are packaged.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.