Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ Nalgene Round LDPE Carboys are the icon of lab water dispensing and storage. Place one at every sink and every work station to enhance your productivity. Carboys have a spigot boss molded in once piece with the carboy for convenience, strength and leak resistance. Suitable for collecting and dispensing distilled water, acids and reagents. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Autoclavable | Not Autoclavable |
Capacity (English) | 2.6 gal. |
Capacity (Metric) | 10 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White (Opaque) |
Description | Round Carboys w/Spigot |
Diameter (Metric) Inner Neck | 64 mm |
Diameter (Metric) Outer | 250 mm |
For Use With (Application) | Collect and dispense distilled water, acids and reagents |
Graduations | Molded-in graduations in 1 gal. and 5 L increments |
Height (Metric) With Closure | 389 mm |
Includes | Integral shoulder handles, Molded-in spigot boss |
Material | LDPE |
Product Type | Carboy with Spigot |
Unit Size | Each |
Autoclavable | Not Autoclavable |
Capacity (English) | 2.6 gal. |
Capacity (Metric) | 10 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White (Opaque) |
Description | Round Carboys w/Spigot |
Diameter (Metric) Inner Neck | 64 mm |
Diameter (Metric) Outer | 250 mm |
For Use With (Application) | Collect and dispense distilled water, acids and reagents |
Graduations | Molded-in graduations in 1 gal. and 5 L increments |
Height (Metric) With Closure | 389 mm |
Includes | Integral shoulder handles, Molded-in spigot boss |
Material | LDPE |
Product Type | Carboy with Spigot |
Unit Size | Case of 6 |
Autoclavable | Not Autoclavable |
Capacity (English) | 1 gal. |
Capacity (Metric) | 4 L |
Closure Material | Polypropylene |
Closure Size | 38-430 |
Color | White |
Description | Round Carboys w/Spigot |
Diameter (Metric) Inner Neck | 26 mm |
Diameter (Metric) Outer | 154 mm |
For Use With (Application) | Collect and dispense distilled water, acids and reagents |
Graduations | Molded-in graduations in 1 gal. and 5 L increments |
Height (Metric) With Closure | 335 mm |
Includes | Integral shoulder handles, Molded-in spigot boss |
Material | LDPE |
No. per Case | 6/Cs. |
Product Type | Carboy with Spigot |
Unit Size | Case of 6 |
Autoclavable | Not Autoclavable |
Capacity (English) | 1 gal. |
Capacity (Metric) | 4 L |
Closure Material | Polypropylene |
Closure Size | 38-430 |
Color | White |
Description | Round Carboys w/Spigot |
Diameter (Metric) Inner Neck | 26 mm |
Diameter (Metric) Outer | 154 mm |
For Use With (Application) | Collect and dispense distilled water, acids and reagents |
Graduations | Molded-in graduations in 1 gal. and 5 L increments |
Height (Metric) With Closure | 335 mm |
Includes | Integral shoulder handles, Molded-in spigot boss |
Material | LDPE |
No. per Case | 6/Cs. |
Product Type | Carboy with Spigot |
Unit Size | Each |
Autoclavable | Not Autoclavable |
Capacity (English) | 6.6 gal. |
Capacity (Metric) | 25 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White (Opaque) |
Description | Round Carboys w/Spigot |
Diameter (Metric) Inner Neck | 64 mm |
Diameter (Metric) Outer | 287 mm |
For Use With (Application) | Collect and dispense distilled water, acids and reagents |
Graduations | Molded-in graduations in 1 gal. and 5 L increments |
Height (Metric) With Closure | 594 mm |
Includes | Integral shoulder handles, Molded-in spigot boss |
Material | LDPE |
Product Type | Carboy with Spigot |
Unit Size | Each |
Autoclavable | Not Autoclavable |
Capacity (English) | 5.3 gal. |
Capacity (Metric) | 20 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White (Opaque) |
Description | Round Carboys w/Spigot |
Diameter (Metric) Inner Neck | 64 mm |
Diameter (Metric) Outer | 286 mm |
For Use With (Application) | Collect and dispense distilled water, acids and reagents |
Graduations | Molded-in graduations in 1 gal. and 5 L increments |
Height (Metric) With Closure | 528 mm |
Includes | Integral shoulder handles, Molded-in spigot boss |
Material | LDPE |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Spigot |
Unit Size | Case of 4 |
Autoclavable | Not Autoclavable |
Capacity (English) | 5.3 gal. |
Capacity (Metric) | 20 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White (Opaque) |
Description | Round Carboys w/Spigot |
Diameter (Metric) Inner Neck | 64 mm |
Diameter (Metric) Outer | 286 mm |
For Use With (Application) | Collect and dispense distilled water, acids and reagents |
Graduations | Molded-in graduations in 1 gal. and 5 L increments |
Height (Metric) With Closure | 528 mm |
Includes | Integral shoulder handles, Molded-in spigot boss |
Material | LDPE |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Spigot |
Unit Size | Each |
Autoclavable | Not Autoclavable |
Capacity (English) | 13 gal. |
Capacity (Metric) | 50 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White (Opaque) |
Description | Round Carboys w/Spigot |
Diameter (Metric) Inner Neck | 64 mm |
Diameter (Metric) Outer | 379 mm |
For Use With (Application) | Collect and dispense distilled water, acids and reagents |
Graduations | Molded-in graduations in 1 gal. and 5 L increments |
Height (Metric) With Closure | 678 mm |
Includes | Integral shoulder handles, Molded-in spigot boss |
Material | LDPE |
Product Type | Carboy with Spigot |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2318-0020PK also known as 2318-0020 | Each | 10 L | 236.10 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2318-0020 | Case of 6 | 10 L | 938.51 Online price 1,200.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2318-0010 | Case of 6 | 4 L | 731.15 Online price 834.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2318-0010PK also known as 2318-0010 | Each | 4 L | 164.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2318-0065 | Each | 25 L | 207.69 Online price 276.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2318-0050 | Case of 4 | 20 L | 691.79 Online price 957.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2318-0050PK | Each | 20 L | 282.30 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2318-0130 | Each | 50 L | 306.38 Online price 415.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Ordering Information:
Supplied with shoulder loop on 1 gal. (3.8L) size, shoulder handles on all other sizes. 1 gal. size is not graduated.
Includes:
Polypropylene spigot and screw closure with TPE gasket † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Nalgene rugged LDPE carboys are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE carboys and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene carboy products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE carboys.