2,3-Dicyanohydroquinone, 98%, Thermo Scientific Chemicals
2,3-Dicyanohydroquinone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dicyanohydroquinone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 232050050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name3,6-dihydroxybenzene-1,2-dicarbonitrile
Molecular FormulaC8H4N2O2
InChI KeyMPAIWVOBMLSHQA-UHFFFAOYSA-N
SMILESOC1=CC=C(O)C(C#N)=C1C#N
Molecular Weight (g/mol)160.13
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Color)Brown to khaki
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only