1x1 image pixel for data collection
1-Butanol, 99.5%, for analysis
1-Butanol, 99.5%, for analysis
1-Butanol, 99.5%, for analysis
1-Butanol, 99.5%, for analysis
Thermo Scientific Chemicals

1-Butanol, 99.5%, for analysis

CAS: 71-36-3 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 232080010
Price (USD)
68.70
Each
In stock
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
68.70
Each
Add to cart
Chemical Identifiers
CAS71-36-3
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Non-volatile matter=<0.001 %
Aldehyde (C4H8O)Passes test
Free acid (C4H8O2)=<0.01 %
Aluminium (Al)=<0.5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link