Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
5 L
Packaging:
View all
Metal Drum
Catalog number 232110250
Price (USD)/ Each
-
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Chemical Identifiers
CAS141-78-6
IUPAC Nameethyl acetate
Molecular FormulaC4H8O2
InChI KeyXEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILESCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Residue after evaporation=<0.001 %
Methyl acetate=<0.1 %
Barium (Ba)=<0.1 ppm
Copper (Cu)=<0.02 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only