Thermo Scientific Chemicals

1-Hydroxypyrene, 99+%, Thermo Scientific Chemicals

Catalog number: 232270500
50 mg, Glass bottle, Each
Thermo Scientific Chemicals

1-Hydroxypyrene, 99+%, Thermo Scientific Chemicals

Catalog number: 232270500
50 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 232270500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5315-79-7
IUPAC Name
pyren-1-ol
Molecular Formula
C16H10O
InChI Key
BIJNHUAPTJVVNQ-UHFFFAOYSA-N
SMILES
OC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Infrared spectrum
Conforms
Appearance (Color)
Beige to brown
Appearance (Form)
Powder or needles
HPLC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text