Thermo Scientific Chemicals

Linalyl acetate, 95%, synthetic, Thermo Scientific Chemicals

Catalog number: 232301000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Linalyl acetate, 95%, synthetic, Thermo Scientific Chemicals

Catalog number: 232301000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 232301000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
115-95-7
IUPAC Name
3,7-dimethylocta-1,6-dien-3-yl acetate
Molecular Formula
C12H20O2
InChI Key
UWKAYLJWKGQEPM-UHFFFAOYNA-N
SMILES
CC(C)=CCCC(C)(OC(C)=O)C=C
Density
(25°C) 0.895 to 0.914 g/cm³
GC
>=94 %
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text