The Pierce Micro BCA Reagent B (MB) is a unique bicinchonic acid (BCA) solution for use in protein concentration determination. This product is sufficient for 3,200 microplate assays when mixed with Reagents MA and MC.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Description
Pierce Micro BCA Reagent B (MB)
Assay
BCA Assay
Detection Method
Colorimetric
Product Line
Micro BCA™, Pierce™
Product Type
Protein Quantitation Assay
Specificity
Not Target-Specific
Sufficient For
3200 Microplate Assays
For Use With (Application)
Solution-based Detection, Absorbance
For Use With (Equipment)
Spectrophotometer, Microplate Reader
Quantity
240 mL
Contents & Storage
Upon receipt store product at room temperature. Product shipped at ambient temperature.
Note: If Reagent MB precipitates upon shipping in cold weather or during long-term storage, dissolve precipitates by gently warming and stirring the solution. Discard any reagent that shows discoloration or evidence of microbial contamination.